Learn More
Cayman Chemical EdrophonIumchlorIde 250mg
SDP
Supplier:  Cayman Chemical 15928250 MG
Encompass_Preferred
A 5-LO inhibitor (IC50 0.8 UM) 100-fold selective for 5-LO over 12-LO and COX inhibits A23187-induced production of 5-HETE and LTB4 (IC50s 0.03 and 0.08 UM respectively) in isolated rat peritoneal macrophages reduces carrageenan-induced paw edema pleurisy and cell infiltration in rats at 80 mg/kg reduces ovalbumin-induced bronchoconstriction in an ovalbumin-sensitized guinea pig model of experimental allergic asthma at 20 mg/kg
Catalog No. 50-296-8751
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.