Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EDTA, 0.0100 Molar, with Magnesium Chloride, Reagents
EDTA, 0.0100 Molar, with Magnesium Chloride, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.
Supplier: Reagents Holdings Llc CE25160020F
Quantity | 20 L |
---|
Chemical Identifiers
7732-18-5, 6381-92-6 | |
18.02 | |
Edetate Disodium, Water Hardness Titrant, (Ethylenedinitrilo)tetraacetic Acid disodium salt | |
O |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water |
Specifications
Colorless | |
100°C | |
Edetate Disodium, Water Hardness Titrant, (Ethylenedinitrilo)tetraacetic Acid disodium salt | |
O | |
18.02 | |
Cubitainer | |
20 L | |
Liquid |
7732-18-5, 6381-92-6 | |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water | |
Reagent | |
1.018 | |
Non-sterile | |
EDTA, 0.0100 Molar, with Magnesium Chloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction