Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDTA, 0.0100 Molar, Reagents

EDTA, 0.0100 Molar, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

$48.25 - $406.00

Chemical Identifiers

CAS 7732-18-5 , 6381-92-6
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Edetate Disodium, Water Hardness Titrant, (Ethylenedinitrilo)tetraacetic Acid disodium salt
IUPAC Name water
SMILES O
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
01-335-359
View Documents
Reagents Holdings Llc
CE25040010F
10 L
Each for $257.00
Add to Cart
 
01-335-358
View Documents
Reagents Holdings Llc
CE2504001A
1 L
Each for $48.25
Add to Cart
 
01-335-357
View Documents
Reagents Holdings Llc
CE25040020F
20 L
Each for $406.00
Add to Cart
 
01-335-356
View Documents
Reagents Holdings Llc
CE2504004A
4 L
Each for $113.60
Add to Cart
 
Description

Description

Specifications

Chemical Identifiers

7732-18-5 , 6381-92-6
18.02
Edetate Disodium, Water Hardness Titrant, (Ethylenedinitrilo)tetraacetic Acid disodium salt
O
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water

Specifications

Colorless
100°C
Edetate Disodium, Water Hardness Titrant, (Ethylenedinitrilo)tetraacetic Acid disodium salt
O
18.02
Cubitainer
4 to 6
Liquid
7732-18-5 , 6381-92-6
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water
Reagent
1.002
Non-sterile
EDTA, 0.0100 Molar
Videos
SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.