Learn More
StatLab EDTA BONE DECALCIFIER BUF 7.4
SDP
Supplier:  StatLab DCV0150
Encompass_Preferred
A bone decalcifier that requires extensive use of special stain techniques. Ideal for iron stains. This decal can take several days to complete the decalcification process.
Catalog No. NC0491492
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.