Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDTA, Disodium Salt, Dihydrate, 0.1M Volumetric Solution, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog No. 02002777 Shop All Avantor Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-002-777 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-002-777 Supplier Avantor Supplier No. 563202

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 6381-92-6
Molecular Formula C10H18N2Na2O10
Molecular Weight (g/mol) 372.24
MDL Number MFCD00150037,MFCD00003541
InChI Key OVBJJZOQPCKUOR-UHFFFAOYSA-L
PubChem CID 44120005
IUPAC Name disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
SMILES O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

Specifications

Packaging Narrow Mouth Polyethylene Bottle
Specific Gravity 1L = 1.02kg
Quantity 1 L
Formula Weight 372.24
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material EDTA, Disodium Salt, Dihydrate, 0.1M Volumetric Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.