Learn More
EDTA Disodium Salt Dihydrate, 99-101.0%, MB Grade, Ultrapure, Thermo Scientific Chemicals
Description
Chelating agent used in many enzyme buffers and as an enzyme inactivator at high concentrations.
Stock Solution (0.5M):
- 186.1 gm EDTA
- H2O to 800 ml
- Titrate with approximately 20 gm of NaOH pellets to pH 8.0.
- Bring solution to 1 liter with water and sterilize by autoclaving.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 248°C (decomposition) |
| Physical Form | Crystalline Powder |
| Quantity | 1 kg |
| Beilstein | 3900609 |
| Formula Weight | 372.24 |
| Percent Purity | 99.0 to 101.0% |
| Grade | Molecular Biology |
| Packaging | Plastic bottle |
| Chemical Name or Material | Ethylenediaminetetraacetic Acid Disodium Salt Dihydrate |
Safety and Handling
| Hazard Category | H332-H373 |
| Precautionary Statement | P260-P271-P304+P340-P312-P314-P501c |
| EINECSNumber | 205-358-3 |
| RTECSNumber | AH4375000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.