Learn More
Research Products International Corp EDTA Free Acid 5 KG

Supplier: Research Products International Corp E581005000.0
Substance Name: EDTA
CAS Number: 60-00-4
Molecular Formula: C10H16N2O8
Molecular Weight: 292.24
Chemical Purity: >99%
Soluble in: Water
Store at: Room Temperature
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.