Learn More
EDTA Solution (0.002 Molar) Volumetric Solution, Reagents
EDTA Solution (0.002 Molar) Volumetric Solution, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier:  Reagents Holdings Llc CE0012001A
Catalog No. 01-335-406
Only null left
Add to Cart
Description
Description
Specifications
Specifications
EDTA Solution (0.002 Molar) Volumetric Solution
7732-18-5, 6381-92-6
C10H14N2O8Na2/H2O
Reagent
1.0001
1 L
Liquid
Colorless
100°C
Edetate Disodium, Water Hardness Titrant, (Ethylenedinitrilo)tetraacetic Acid disodium salt
Poly Bottle
4 to 6
Non-sterile
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.