Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDTA Tetrasodium Salt, Dihydrate, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog No. 02002779 Shop All Avantor Products
Change view
Click to view available options
Quantity:
2.5 kg
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-002-779 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 02-002-779 Supplier Avantor Supplier No. L69309

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

    For Laboratory,Research,or Manufacturing Use

Chemical Identifiers

CAS 10378-23-1
Molecular Formula C10H20N2Na4O10
Molecular Weight (g/mol) 420.233
InChI Key DGWOWDJDIBPYOR-UHFFFAOYSA-N
Synonym ethylenediaminetetraacetic acid tetrasodium salt dihydrate, tetrasodium ethylenediaminetetraacetate dihydrate, edta-na4, edta tetrasodium salt, edta tetrasodium salt hydrate, ethylenediaminetetraacetic acid tetrasodium salt hydrate, 64-02-8 anhydrous, ksc909a7h, edta tetrasodium salt dihydrate, tetrapotassium ion 4-edta dihydrate
PubChem CID 131843362
IUPAC Name 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid;sodium;dihydrate
SMILES C(CN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O.O.O.[Na].[Na].[Na].[Na]

Specifications

Packaging Poly Bottle
Quantity 2.5 kg
Specific Gravity 1L = 0.71kg
Formula Weight 416.21
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material EDTA Tetrasodium Salt, Dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.