Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDTA Titrant, 0.00100 M (M/1000), 1 mL = 0.1 mg CaCO3 (0.04 mg Ca), Ricca Chemical
SDP

Catalog No. 269216 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
2692-16 500 mL Natural Poly Bottle
2692-32 1 L Natural Poly Bottle
R26920004A 4 L Natural Poly Bottle
R269200010F 10 L Cubitainer
R269200020F 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 2692-16 Supplier Ricca Chemical Company Supplier No. 269216
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6381-92-6 , 7732-18-5
Molecular Formula C10H18N2Na2O10
Molecular Weight (g/mol) 372.24
MDL Number MFCD00150037,MFCD00003541
InChI Key OVBJJZOQPCKUOR-UHFFFAOYSA-L
Synonym edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
PubChem CID 44120005
IUPAC Name disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
SMILES O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

Specifications

Color Colorless
CAS Min % 98.0, 0.04
CAS Max % 100.0, 0.04
Equivalence 1 mL = 0.1 mg CaCO
Grade Laboratory
Packaging Natural Poly Bottle
pH 4 to 6
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 0.00100Molar
Chemical Name or Material EDTA Titrant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.