Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDTA Titrant, 0.0500 M (M/20), Ricca Chemical
SDP

Catalog No. p-3251635 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
2705-16 500 mL Natural Poly Bottle
2705-32 1 L Natural Poly Bottle
2705-1 4 L Natural Poly Bottle
2705-2-5 10 L Cubitainer
2705-5 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 2705-16 Supplier Ricca Chemical Company Supplier No. 270516
Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 6381-92-6 , 7732-18-5
Molecular Formula C10H18N2Na2O10
Molecular Weight (g/mol) 372.24
MDL Number MFCD00150037,MFCD00003541
InChI Key OVBJJZOQPCKUOR-UHFFFAOYSA-L
Synonym edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
PubChem CID 44120005
IUPAC Name disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
SMILES O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

Specifications

Color Colorless
CAS Min % 96.0, 1.79
CAS Max % 100.0, 1.86
Grade Laboratory
Packaging Natural Poly Bottle
pH 4 to 6
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 0.0500Molar
Chemical Name or Material EDTA Titrant
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.