Learn More
AquaPhoenix Scientific™ Education Aquarium Tanks
SCI_ED
Catalog No. S06524 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Capacity (English):
10 gal.
20 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
S06524 20 gal.
S06522 10 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S06524 Supplier AquaPhoenix Scientific™ Supplier No. AQ5250G
Add to Cart
Add to Cart
Tanks in sizes from 10 to 55 gallons.
  • Aquariums are made with glass panels, fitted and silicone-sealed into injected molded frames
  • Lighted hood covers and electronic ballast hold fixtures for a single fluorescent bulb (available separately)
Specifications
Capacity (English) 20 gal.
Material Glass