Learn More
Bachem N-Hydroxysuccinimide
SDP
Supplier:  Bachem 4025878.0500
Encompass_Preferred
N-Hydroxysuccinimide, 6066-82-6, C4H5NO3, Mr 115.09, 500g. HOSu. Educt for the synthesis of N-hydroxysuccinimide esters of N-protected amino acids and peptides. Additive for peptide synthesis by the azide fragment condensation on a soluble polymer support. Chiral stabilizer for peptide coupling reactions.
Catalog No. 50-194-7383
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.