Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Investigating Synthetic Biology
SDPSCI_ED

Catalog No. S66821 Shop All EDVOTEK Products
Change view
Click to view available options
Detection Method:
Gel Electrophoresis
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detection Method
S66821 Gel Electrophoresis
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S66821 Supplier Edvotek Supplier No. 331
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

PCR EdvoBeads™, primer mix concentrate, template DNA concentrate, universal DNA buffer, ultrapure water, DNA standard marker, 10X gel loading solution, purification and wash buffers, spin columns and reservoirs, Dryzyme™ mix, Dryzyme™ reconstitution buffer, enzyme reaction buffer, vector cut with EcoRI and HindIII, T4 DNA ligase, ReadyPour LB agar, LB medium, BactoBeads™ (Host), Calcium Chloride, competent cell solution, control DNA, recovery broth, Kanamycin, IPTG, Salacylic acid, wintergreen scent, 50X electrophoresis buffer, UltraSpec-Agarose™, SYBR™ Safe Stain, microcentrifuge and PCR tubes, 10 ml pipet, 50 ml conical tubes, sterile loops

Requires

  • Horizontal gel electrophoresis apparatus
  • Thermal cycler
  • Water baths
  • Incubation oven
  • Hotplate
  • Micropipets
  • Balance
  • UV transilluminator
  • Microtest tubes
  • 250mL flasks
  • Ice
  • All necessary safety equipment
  • Distilled or deionized water

Cloning experiment for the classroom.

Teach your students about synthetic biology with this exciting and exclusive lab! Students use PCR to amplify the coding sequence of the BSMT1 enzyme. This interesting enzyme is responsible for the formation of methyl salicylate, a chemical with a strong 'wintergreen' odor. The PCR product is purified, restriction digested, and inserted into a plasmid vector. The resulting recombinant DNA is then used to transform E. coli BactoBeads™. Finally, students design an experiment to express the enzyme from their transformants and perform a smell test to confirm that the bacterial factories are working!

Specifications

Content And Storage Instructions, biologicals, Buffers and reagents for PCR, ligation and transformation, ReadyPour™ luria broth agar, DNA size ladder, wax beads, Agarose, electrophoresis Buffer, Gel stain. Store at -20°C
Detection Method Gel Electrophoresis
Form Gel
GC-Rich PCR Performance Medium
PCR Method Standard PCR
Reaction Speed 30 to 90 min.
Label or Dye SYBR Green, FlashBlue Stain
Product Type Classroom Experiment
Quantity 1 Kit
For Use With (Application) Standard PCR
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.