Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ DNA/RNA Microarrays
SDPSCI_ED

Catalog No. s05648 Shop All EDVOTEK Products
Change view
Click to view available options
Dimensions (LxW):
2 x 4 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (LxW)
S05648 2 x 4 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S05648 Supplier Edvotek™ Supplier No. 235
Add to Cart
Add to Cart

Includes

  • Instructions
  • Simulated patient DNA and RNA samples and controls
  • Microarray cards
  • Plastic bags to incubate membrane
  • Microtest tubes
  • Pipets

Requires

Automatic micropipets and tips, distilled water and beakers or flasks

Membrane microarray technology enables scientists to screen large numbers of samples in one assay.

Benefit from cost savings by reducing the sample size, while saving time and yielding accurate results.

  • Students apply simulated DNA and RNA samples to a membrane to screen for positive and negative samples
  • Complete in one hour

Order Info

For 10 groups.

Specifications

Coating Uncoated
Thickness (Metric) 1 mm
Includes Instructions, simulated patient dna and rna samples, controls, microarray cards, plastic bags to incubate membrane, microtest tubes, pipets
Species Human
Tissue Condition Normal
Type Dna or Rna Microarray Kit
Dimensions (LxW) 2 x 4 in.
Enhanced Surface No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.