Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Drosophila Genotyping Using PCR
SDPSCI_ED

Catalog No. S66827 Shop All EDVOTEK Products
Change view
Click to view available options
Detection Method:
Gel Electrophoresis
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detection Method
S66827 Gel Electrophoresis
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S66827 Supplier Edvotek Supplier No. 337
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Flies
  • Primers
  • Buffers
  • Chemicals
  • Instructions and background information
  • Study questions

Requires

  • Horizontal gel electrophoresis apparatus
  • Thermal cycler
  • Hotplate
  • Micropipetter
  • Balance
  • Water bath
  • Microcentrifuge
  • 500mL flasks
  • UV transilluminator
  • Pipet pump or bulb
  • Isopropanol
  • All necessary safety equipment
  • Distilled or deionized water

Advanced-level experiment uses agarose gel electrophoresis to explore DNA polymorphisms.

Students will learn about DNA polymorphisms by amplifying DNA regions that vary between wild & mutant Drosophila. Amplified DNA from wild-type and white-eyed flies are separated by agarose gel electrophoresis and analyzed.

Order Info

Class size: 10 groups (20 PCR reactions).

Specifications

Content And Storage Instructions, PCR EdvoBeads™, primers, DNA extraction Buffer, 200bp ladder, proteinase K, drosophila (wild type and white eye), Agarose, electrophoresis Buffer, InstaStain™ Ethidium Bromide and FlashBlue™ Stain. Store at -20°C
Detection Method Gel Electrophoresis
Form Lyophilized
GC-Rich PCR Performance Medium
PCR Method Standard PCR
Reaction Speed 30 to 90 min.
Label or Dye SYBR Green, FlashBlue Stain
Product Type Classroom Experiment
Quantity 1 Kit
For Use With (Application) Genotyping
No. of Reactions 20
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.