Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Identification of Genetically Modified Foods Using PCR
SDPSCI_ED

Catalog No. s68638 Shop All EDVOTEK Products
Change view
Click to view available options
Detection Method:
Gel Electrophoresis
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detection Method
S68638 Gel Electrophoresis
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S68638 Supplier Edvotek Supplier No. 962
Add to Cart
Add to Cart

Includes

Instructions, PCR EdvoBeads™, Universal DNA Buffer, TE Buffer, LyphoPrimer™ Mix, GMO Positive LyphoControl™, NaCl Solution, DNA Extraction Buffer, 100 base pair ladder, Proteinase K, UltraSpec-Agarose™, TBE Electrophoresis Buffer Powder, SYBR™ Safe Stain, 0.2 mL PCR Tubes, Plastic Micropestles, Transfer Pipets, and Snap-top Microcentrifuge Tubes

Requires

  • Thermal cycler or three water baths
  • Horizontal gel apparatus
  • DC power supply
  • Microcentrifuge
  • Balance
  • Microwave oven or hotplate
  • UV transilluminator
  • Pipetter

Certain foodstuffs, such as tofu and soy milk, contain raw materials from genetically modified organisms (GMOs).

Some foods contain raw materials from genetically modified organisms (GMO). Examples include tofu, corn flakes and corn meal. In this experiment, your students will extract DNA from food or plant material and perform PCR to determine if any GM indicator genes are present. Amplified DNA is separated and sized by agarose gel electrophoresis.

Order Info

Class size: 10 groups.

Specifications

Content And Storage Instructions, DNA extraction reagents, PCR EdvoBeads™, microtubes, primers, DNA ladder, wax beads, Gel loading dye, Agarose, electrophoresis Buffer, InstaStain™ Ethidium Bromide and FlashBlue™ Stain. Store at -20°C
Detection Method Gel Electrophoresis
Form Lyophilized
GC-Rich PCR Performance Medium
PCR Method Standard PCR
Reaction Speed 30 to 90 min.
Label or Dye SYBR Green, FlashBlue Stain
Product Type Classroom Experiment
Quantity 1 Kit
For Use With (Application) Food and Beverage
No. of Reactions 25
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.