Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Edvotek™ M36 HexaGel Electrophoresis Apparatus
SDPSCI_ED

Catalog No. S11515A Shop All EDVOTEK Products
Change view
Click to view available options
Capacity:
6 Gel Trays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
S11515A 6 Gel Trays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S11515A Supplier Edvotek™ Supplier No. 515
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Apparatus allows you to run six gels and separate up to 36 samples simultaneously.

Six groups of students can load their own individual gels and six gels are run together for 30-40 minutes giving results.Eliminate cumbersome gel tray taping and pour gels quickly and easily with our innovative gel tray sealing rubber end caps.

  • New, sleek design
  • Contoured lid enhances visualization
  • Vented base
  • Color coded push tabs
  • Pour spout to pour out used buffer
  • Replaceable electrode modules

Specifications

Capacity 6 Gel Trays
Detection Method Seperation
For Use With (Application) Gel Electrophoresis
Gel Size Mini
Max. Current 500 mA
Running Time 15 to 30 min.
Timer 0 to 99 hr. or continuous
Type Horizontal Electrophoresis System
Includes Six 7 x 7cm trays, six 6-teeth combs, twelve Rubber end caps
Model M36 HexaGel
Volume (English) Buffer 33.8 oz.
Volume (Metric) Buffer 1000 mL
Warranty Lifetime
Max. Voltage 200 V
Voltage 75/150 V
Frequency 50/60 Hz
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.