Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Micropipet Tips
SDPSCI_ED

Catalog No. S06689 Shop All EDVOTEK Products
Change view
Click to view available options
Volume (Metric):
0.5 to 10 μL
1 to 200 μL
1000 to 5000 μL
200 to 1000 μL
Format:
Bagged
Rack
Rack, 2 x 100
Rack, 2 x 96
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Format
S06689 1 to 200 μL Rack, 2 x 96
S06688 0.5 to 10 μL Bagged
S06691 200 to 1000 μL Rack, 2 x 100
S06690 1 to 200 μL Bagged
S06687 0.5 to 10 μL Rack, 2 x 96
S06692 200 to 1000 μL Bagged
S06693 1000 to 5000 μL Bagged
S06694 1 to 200 μL Rack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
CHOKING HAZARD - Small Parts. Not for children under 6 years.
Catalog No. S06689 Supplier Edvotek™ Supplier No. 636
Add to Cart
Add to Cart
CHOKING HAZARD - Small Parts. Not for children under 6 years.

Available in a variety of volumes and formats

  • Includes: Ultra, Yellow, Blue, Fine tip and Jumbo micropipet tips

Specifications

Volume (Metric) 1 to 200 μL
Filtered Non-filtered
Sterility Non-sterile
Color Yellow
Autoclavable Autoclavable
Format Rack, 2 x 96
Tip Style Standard
Product Type Pipette Tip
Disposable Limited-use
For Use With (Equipment) Universal Fit
Graduated No
Material Plastic
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.