Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Mitochondrial DNA Analysis Using PCR
SDPSCI_ED

Catalog No. S66822 Shop All EDVOTEK Products
Change view
Click to view available options
Detection Method:
Gel Electrophoresis
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detection Method
S66822 Gel Electrophoresis
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S66822 Supplier Edvotek Supplier No. 332
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Instructions, PCR EdvoBeads™, Mitochondrial Primer Mix Concentrate, EdvoQuick™ DNA Ladder, Control DNA Concentrate, TE Buffer, Proteinase K, UltraSpec-Agarose™, Electrophoresis Buffer (50X), 10X Gel Loading Solution, InstaStain™ Ethidium Bromide, FlashBlue™ Stain, Microcentrifuge & PCR Tubes, Disposable Plastic Cups, Salt Packets, & Wax Beads

Requires

  • Thermal cycler
  • Horizontal gel electrophoresis apparatus
  • Microcentrifuge
  • Hotplate
  • Micropipets
  • Balance
  • Water bath
  • UV transilluminator
  • 500mL flasks
  • All necessary safety equipment
  • Distilled or deionized water

Advanced-level experiment uses the Polymerase Chain Reaction (PCR) to amplify a specific region of the mitochondrial chromosome and perform gel electrophoresis.

  • Energy within the cell is produced by the organelles called mitochondria, which have their own DNA (mtDNA)
  • Mitochondrial diseases have been mapped to the mtDNA chromosome
  • Forensic identification also commonly examines mtDNA

Order Info

Contains materials for 25 individual reactions and six gels.

Specifications

Content And Storage Instructions, proteinase K, PCR EdvoBeads™, control DNA and primers, microtubes, chelating agent, Agarose, DNA ladder, practice Gel loading solution, Gel loading dye, Buffer, InstaStain™ Ethidium Bromide and FlashBlue™ Stain. Store at -20°C
Detection Method Gel Electrophoresis
Form Lyophilized
GC-Rich PCR Performance Medium
PCR Method Standard PCR
Reaction Speed 30 to 90 min.
Label or Dye SYBR Green, FlashBlue Stain
Product Type Classroom Experiment
Quantity 1 Kit
For Use With (Application) Standard PCR
No. of Reactions 25
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.