Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ PCR-Based DNA Typing
SDPSCI_ED

Catalog No. S66824 Shop All EDVOTEK Products
Change view
Click to view available options
Content And Storage:
Instructions, proteinase K, PCR EdvoBeads™, control DNA and primers, microtubes, chelating agent, Agarose, DNA ladder, practice Gel loading solution, Gel loading dye, Buffer, FlashBlue™ stain. Store at -20°C
Instructions, proteinase K, PCR EdvoBeads™, control DNA and primers, microtubes, chelating agent, Agarose, DNA ladder, practice Gel loading solution, Gel loading dye, Buffer, InstaStain™ Ethidium Bromide and FlashBlue™ Stain. Store at -20°C
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Content And Storage
S66824 Instructions, proteinase K, PCR EdvoBeads™, control DNA and primers, microtubes, chelating agent, Agarose, DNA ladder, practice Gel loading solution, Gel loading dye, Buffer, FlashBlue™ stain. Store at -20°C
S66823 Instructions, proteinase K, PCR EdvoBeads™, control DNA and primers, microtubes, chelating agent, Agarose, DNA ladder, practice Gel loading solution, Gel loading dye, Buffer, InstaStain™ Ethidium Bromide and FlashBlue™ Stain. Store at -20°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S66824 Supplier Edvotek Supplier No. 334
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Instructions, PCR EdvoBeads™, D1S80 Primer Mix Concentrate, EdvoQuick™ DNA Ladder, Control DNA Concentrate, TE Buffer, Proteinase K, UltraSpec-Agarose™, TBE Electrophoresis Buffer Powder, 10x Gel Loading Solution, InstaStain™ Ethidium Bromide, Microcentrifuge & PCR Tubes, Disposable Plastic Cups, Salt Packets, & Wax Beads

Requires

  • Thermal cycler
  • Horizontal gel electrophoresis apparatus
  • Microcentrifuge
  • Automatic micropipets
  • UV transilluminator
  • 500mL flasks
  • Glassware
  • All necessary safety equipment
  • Ice
  • Ice bucket
  • Distilled or deionized water

DNA typing adapted from FBI technology.

Safe experiment for classroom use to demonstrate DNA fingerprinting.

Alu-Human DNA Typing (S66823)

  • Examine students' DNA for an Alu element on Chromosome 16
  • Investigate DNA patterns
  • Compare Alu genotypes with those of their classmates

VNTR Human DNA Typing (S66824)

  • Use PCR to fingerprint students' DNA at the D1S80 locus on Chromosome 1
  • Provide an example of the variable number of tandem repeat (VNTR) polymorphism

Specifications

Content And Storage Instructions, proteinase K, PCR EdvoBeads™, control DNA and primers, microtubes, chelating agent, Agarose, DNA ladder, practice Gel loading solution, Gel loading dye, Buffer, FlashBlue™ stain. Store at -20°C
Detection Method Gel Electrophoresis
Form Lyophilized
GC-Rich PCR Performance Medium
PCR Method Standard PCR
Reaction Speed 60 min.
Label or Dye SYBR Green, FlashBlue Stain
Product Type Classroom Experiment
Quantity 1 Kit
For Use With (Application) Standard PCR
No. of Reactions 25
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.