Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Proteinase K
SDPSCI_ED

Catalog No. S05681 Shop All EDVOTEK Products
Change view
Click to view available options
Quantity:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S05681 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S05681 Supplier Edvotek™ Supplier No. 626
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Required to prepare the lysis solution for isolation of DNA from hair.

  • Proteinase K is required to prepare the lysis solution for isolation of DNA from hair

Specifications

Concentration 10,000X
For Use With (Application) Preparing lysis solution for isolation of DNA from hair
Sensitive to Heat Inactivation Yes
Source Hair
Quantity Each
Product Type Proteinase K
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.