Learn More
MBL International Corporation Anti-EEA1 mAb (Monoclonal Antibody)
SDP
Supplier:  MBL International Corporation M1763
Encompass_Preferred
Clonality:Monoclonal; Application:ICC|IP|WB; Conjugate:Unlabeled; Species Reactivity:Human|Mouse|Rat; Size:100 µl; Target:EEA1; Monoclonal Antibody of 100 µl targeting EEA1 for ICC, IP, WB.EEA1 Monoclonal Antibody.
Catalog No. 50-506-16
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.