Learn More
Selleck Chemical LLC Efonidipine S4977-25mg
SDP
Supplier:  Selleck Chemical LLC S497725MG
Encompass_Preferred
Efonidipine (NZ-105) is an L- and T-type calcium channel blocker leading to vasodilation and decreased automaticity of the heart It also suppresses aldosterone secretion from the adrenal
Catalog No. 50-310-9004
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.