Learn More
Selleck Chemical LLC Tomivosertib (eFT-508) 25mg 1849590-01-7
SDP
Supplier:  Selleck Chemical LLC S827525MG
Encompass_Preferred
Tomivosertib (eFT-508) is a potent and selective MNK1/2 inhibitor with IC50s of 2.4 nM and 1 nM, respectively. It potentially results in decreased tumor cell proliferation and tumor growth. Tomivosertib (eFT-508) inhibits eIF4E phosphorylation and dramatically downregulates PD-L1 protein abundance. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5881
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.