Learn More
Enzo Life Sciences (+)-Egenine (25mg). CAS: 6883-44-9
SDP
Supplier:  Enzo Life Sciences ALX350362M025
Encompass_Preferred
First phthalideisoquinoline hemiacetal from a natural source. Inhibits Ca2+ currents and contractile responses in vitro. Alternative name: 6-(6-Methyl-5,6,7,8-tetrahydro[1,3]dioxolo[4,5-g]isoquinolin-5-yl)-6,8-dihydrofuro[3,4-E][1,3]benzodioxol-8-ol, (+)-Decumbensine. Purity: ≥98% (HPLC). Solubility: Soluble in ethyl acetate or chloroform; sparingly soluble in 100% ethanol or ether; practically insoluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-8862
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.