Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EGF Polyclonal Antibody, PeproTech®, Invitrogen™

Catalog No. p-7245544
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
50 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
500P174G50 50 μg
500P174G100 100 μg
500P174G1MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 500P174G50 Supplier Invitrogen™ Supplier No. 500P174G50UG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Goat Polyclonal Antibody

AA Sequence of recombinant protein: NSYPGCPSSY DGYCLNGGVC MHIESLDSYT CNCVIGYSGD RCQTRDLRWW ELR. Preparation: Produced from sera of goats immunized with highly pure Recombinant Murine EGF. Anti-Murine EGF-specific antibody was purified by affinity chromatography employing an immobilized Murine EGF matrix. Sandwich ELISA: To detect Murine EGF by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Murine EGF (500-P174GBt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Murine EGF. Western Blot: To detect Murine EGF by Western Blot analysis this antibody can be used at a concentration of 0.1- 0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Murine EGF is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

EGF (Epidermal growth factor) exerts its actions by binding to the EGF Receptor, a 170 kDa protein kinase. Activation of EGFR initiates diverse cellular pathways in response to toxic environmental stimuli, or to EGF binding to the receptor, the EGFR forms homo- or heterodimers with other family members. Each dimeric receptor complex initiates a distinct signaling pathway by recruiting different Src homology 2 (SH2) containing effector proteins. EGF is far and wide expressed in kidney, cerebrum, prostrate and salivary glands. EGF acts as a potent mitogenic factor and the phosphorylated receptor recruits adapter proteins like GRB2 that activates complex downstream signaling cascades. EGF activates at least 4 major downstream signaling cascades including the RAS-RAF-MEK-ERK, PI3 kinase-AKT, PLCgamma-PKC and STAT modules. Research studies suggest the protein may also play important role in activating the NF-kappa-B signaling cascade. Defects in the EGGF gene are the cause of hypomagnesemia type 4 and dysregulation has been associated with the growth and progression of certain cancers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EGF
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene EGF
Gene Accession No. P01132
Gene Alias AI790464; beta-urogastrone; EGF; Epidermal growth factor; epidermal growth factor (beta-urogastrone); epidermal growth factor precursor; H-EGF; HOMG4; Pro-epidermal growth factor; Pro-epidermal growth factor precursor (EGF); sb:eu639; URG; Urogastrone
Gene Symbols EGF
Host Species Goat
Immunogen E.coli-derived Recombinant Murine EGF.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13645
Target Species Mouse
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.