Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EGFL4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537453
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537453 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537453 Supplier Invitrogen™ Supplier No. PA537453
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 255 kDa. Purity is >95% by SDS-PAGE.

EGFL4 (epidermal growth factor-like protein 4), also known as MEGF8 (multiple epidermal growth factor-like domains protein 8) or C19orf49, is a 2,845 amino acid single-pass type I membrane protein. EGFL4 contains 2 CUB domains, 5 EGF-like domains, 12 Kelch repeats, 4 laminin EGF-like domains and 7 PSI domains. Existing as two alternatively spliced isoforms, the gene encoding EGFL4 maps to human chromosome 19q13.2. Chromosome 19 consists of approximately 63 million bases, makes up over 2% of human genomic DNA, and is recognized for having the greatest gene density of the human chromosomes. It is the genetic home for a number of immunoglobulin superfamily members including the killer cell and leukocyte Ig-like receptors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EGFL4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene MEGF8
Gene Accession No. P60882, Q7Z7M0, Q9QYP0
Gene Alias AW049492; b2b1702Clo; b2b288Clo; C19orf49; CRPT2; EGFL4; EGF-like domain-containing protein 4; EGF-like protein 4; EGF-like-domain, multiple 4; Epidermal growth factor-like protein 4; HBV pre-s2 binding protein 1; HBV pre-S2-binding protein 1; hepatitis B virus pre-S2-binding protein 1; KIAA0817; m687Ddg; Megf8; mKIAA0817; multiple EGF like domains 8; multiple EGF-like domain protein 4; multiple EGF-like domains protein 8; multiple EGF-like-domains 8; multiple epidermal growth factor-like domains protein 8; SBP1
Gene Symbols MEGF8
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 1245-1296 of human EGFL4.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114029, 1954, 269878
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.