Learn More
Invitrogen™ EGFL7 Monoclonal Antibody (OTI5C9)

Description
EGFL7 Monoclonal Antibody for IHC (P)
Specifications
Specifications
| Antigen | EGFL7 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | OTI5C9 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | EGFL7 |
| Gene Accession No. | Q9UHF1 |
| Gene Alias | Cbl20; EGF like domain multiple 7; EGFL7; EGF-like domain 7; EGF-like domain-containing protein 7; EGF-like protein 7; EGF-like-domain, multiple 7; Epidermal growth factor-like protein 7; Estrogen-regulated protein CBL20 20.4kD; Estrogen-regulated protein CBL20, 20.4kD; Megf7; multiple EGF-like domain protein 7; multiple EGF-like domains protein 7; multiple epidermal growth factor-like domain protein 7; multiple epidermal growth factor-like domains protein 7; NEU1; NEU1 protein; NOTCH4-like protein; RP11-251M1.2; UNQ187/PRO1449; Vascular endothelial statin; vascular endothelial-statin; VE-STATIN; Zneu1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.