Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EGFL7 Monoclonal Antibody (OTI5C9), TrueMAB™, OriGene
SDP

Catalog No. 501681810
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-1810 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-1810 Supplier OriGene Supplier No. TA807863
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Epidermal growth factor-like protein 7 (EGFL7), also known as VE-statin, MEGF7, Notch4-like protein or Zneu1, is a 29.6 kDa protein produced and secreted by endothelium. The protein is also expressed by some normal and maligt tissues. The main function of EGFL7 is to regulate angiogenesis and vasculogenesis. A high level of EFGL-7 can be found in tissues with active angiogenesis of a variety of disorders.

Specifications

Antigen EGFL7
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI5C9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene EGFL7
Gene Accession No. Q9UHF1
Gene Alias NEU1, VE-STATIN, ZNEU1
Gene Symbols EGFL7
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 24-273 of human EGFL7 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51162
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.