Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eGFP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PICAB4211 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PICAB4211 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PICAB4211 Supplier Invitrogen™ Supplier No. CAB4211
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

CAB4211 detects eGFP in samples. CAB4211 has been successfully used in Western Blot and ICC/IF applications. The CAB4211 immunogen is purified recombinant eGFP.

eGFP (green fluorescent protein) fluorescent protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen eGFP
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation BBS with no preservative; pH 8.4
Gene Alias eGFP; egfp tag; GFP; GFP tag; GFP2; green fluorescence; green fluorescent; Green Fluoresecent Protein; Turbo eGFP
Host Species Rabbit
Immunogen Purified recombinant eGFP.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Tag
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.