Learn More
ACROBiosystems ACROBiosystems Human EGF R Protein, His Tag, low endotoxin
Supplier: ACROBiosystems EGRH522A100UG
Recombinant Protein;100UG;Human EGF R, His Tag, low endotoxin (EGR-H522a) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 645 (Accession # P00533-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 70.5kDa. The protein migrates as 110-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cetuximab at 2 μg/mL (100 μL/well) can bind Human EGF R, His Tag, low endotoxin (Cat. No. EGR-H522a) with a linear range of 0.8-6 ng/mL (QC tested).;Immobilized Erbitux on CM5 Chip via anti-human Fc IgG, can bind Human EGF R, His Tag, low endotoxin (Cat.No. EGR-H522a) with an affinity constant of 0.492 nM as determined in SPR assay (Biacore T200) ( Routinely tested).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.