Learn More
Abcam EGFR Peptide, Synthetic, Mouse, >90% Purity, 1 mg
Description
- Target Protein: EGFR
- Type: Synthetic
- Purity: >90%
- Form: Liquid
- Tested Application: BL
- Formulation: HEPES, 0.87% Sodium chloride, 0.0584% EDTA, 0.001% Sorbitan monolaurate, ethoxylated
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | EGFR (Mouse) |
| Substrate Properties | Peptide-based Substrate |
| Shipping Condition | Blue Ice |
| Reactivity | Mouse |
| Content And Storage | Store at -20°C. |
| Buffer | HEPES, 0.87% sodium chloride, 0.0584% EDTA, 0.001% sorbitan monolaurate, ethoxylated |
| Concentration | 1 mg/mL |
| Form | Liquid |
| Label or Dye | Unconjugated |
| Cross Reactivity | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.