Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Egg Shaped Stir Bar
SDPSCI_ED

Catalog No. S99480 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Length (Metric):
15 mm
30 mm
50 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
S99480 15 mm
S99482 30 mm
S99483 50 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S99480 Supplier United Scientific™ Supplier No. MSHL15
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

United Scientific Egg Shaped Stir Bar for use with Hot Plate/Stirrers.

  • Egg-shaped magnetic stir bar with blunt edges
  • High-quality magnet coated with PTFE
  • Available in multiple sizes and designs

Specifications

Material Alnico PTFE
Quantity 1/Pk.
Color White
Shape Egg
Type Stir Bar
Diameter (Metric) 7 mm
Length (Metric) 15 mm
Autoclavable Autoclavable
Coating PTFE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.