Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ EGLN1/PHD2 Recombinant Protein Antigen
SDP

Catalog No. NBP254676X Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP254676X 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP254676X Supplier Novus Biologicals™ Supplier No. NBP254676PEP
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human EGLN1/PHD2 The EGLN1/PHD2 Recombinant Protein Antigen is derived from E. coli. The EGLN1/PHD2 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.

Specifications

Purity >80% by SDS-PAGE and Coomassie blue staining
Common Name EGLN1 Recombinant Protein Antigen
Content And Storage Store at −20°C long term. Avoid freeze-thaw cycles
Formulation In PBS (pH 7.4) containing 10% glycerol
For Use With (Application) AC
Gene Alias C1orf12, C1orf12HPH2
Gene Symbol EGLN1
Label Type Unlabeled
Molecular Weight (g/mol) 27 kDa
Product Type Recombinant Protein Antigen
Quantity 0.1 mL
Regulatory Status RUO
Source E.coli
Specific Reactivity This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-53727. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.