Learn More
Lonza Walkersville EGM-2 Endothelial Cell Growth Medium-2 BulletKit
Supplier: Lonza Walkersville CC3162
Choose from a variety of endothelial cell growth media for an easy cultivation of primary endothelial cells. Every endothelial cell growth medium has its special formulation and is optimized to grow certain cell types. EGM - 2 Medium is suitable to grow most types of large vessel endothelial cells. Each type of endothelial cell growth medium is guaranteed to perform when used to grow Lonza's Endothelial Primary Cells.Content:1 x EBM-2 Basal Medium (CC-3156), 500 mL1 x EGM-2 SingleQuots Supplement Pack (CC-4176) containing:1 x Bottle FBS, 10.00 mL1 x Natural Cap Vial with Hydrocortisone, 0.20 mL1 x Gray Cap Vial with hFGF-B, 2.00 mL1 x White Cap Vial with VEGF, 0.50 mL1 x Yellow Cap Vial with R3-IGF-1, 0.50 mL1 x Blue Cap Vial with Ascorbic Acid, 0.50 mL1 x Green Cap Vial with hEGF, 0.50 mL1 x Red Cap Vial with GA-1000, 0.50 mL1 x Orange Cap Vial with Heparin, 0.50 mL