Learn More
Lonza Walkersville EGM-2 MV Microvascular Endothelial SingleQuots Kit
Supplier:  Lonza Walkersville CC4147
Encompass
EGM -2 MV SingleQuot Kit are components sold separately and does not include the basal medium. These are part of the EGM -2 MV Microvascular Endothelial Cell Growth Medium BulletKit (Catalog Number CC-3202) which consists of basal medium and supplements packaged together. Choose from a variety of endothelial cell growth media for an easy cultivation of primary endothelial cells. Every endothelial cell growth medium has its special formulation and is optimized to grow certain cell types. Each type of endothelial cell growth medium is guaranteed to perform when used to grow Clonetics Primary Cells. Content:1 x Bottle FBS 25.00 mL1 x Natural Cap Vial with Hydrocortisone 0.20 mL1 x Gray Cap Vial with hFGF-B 2.00 mL1 x White Cap Vial with VEGF 0.50 mL1 x Yellow Cap Vial with R3-IGF-1 0.50 mL1 x Blue Cap Vial with Ascorbic Acid 0.50 mL1 x Green Cap Vial with hEGF 0.50 mL1 x Red Cap Vial with GA-1000 0.50 mL
Catalog No. NC9729653