Learn More
Alpco Ltd EHEC Verotoxin ELISA
SDP
Supplier:  Alpco Ltd 86VTXHUE01
Encompass
The Verotoxin ELISA is an enzyme immunoassay for the qualitative detection of verotoxins (syn. shiga toxins) from a stool enrichment in mTSB broth (sold separately, see Enrichment Broth catalog# 86-VTXBR-100). For Research Use Only. Not for use in diagnostic procedures.
Catalog No. NC2885457
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.