Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EHMT2 Mouse anti-Human, Mouse, Clone: 1A12E3, Proteintech
SDP

Catalog No. 501737106 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-7106 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-7106 Supplier Proteintech Group Inc Supplier No. 666891IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This gene encodes an enzyme, consisting of two identical proteins, which catalyzes the isomerization of glyceraldehydes 3-phosphate and dihydroxy-acetone phosphate in glycolysis and gluconeogenesis. Mutations in this gene are associated with triosephosphate isomerase deficiency. Pseudogenes have been identified on chromosomes 1, 4, 6 and 7. Alternative splicing results in multiple transcript variants.

Specifications

Antigen EHMT2
Applications Western Blot
Classification Monoclonal
Clone 1A12E3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene EHMT2
Gene Accession No. Q96KQ7, Q9Z148
Gene Alias BAT8, C6orf30, DKFZp686H08213, EHMT2, FLJ35547, G9A, H3 K9 HMTase 3, HLA B associated transcript 8, KMT1C, Lysine N methyltransferase 1C, NG36, NG36/G9a, Protein G9a
Gene Symbols EHMT2
Host Species Mouse
Immunogen EHMT2 Fusion Protein Ag27477
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10919, 110147
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.