Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EHMT2 Recombinant Rabbit Monoclonal Antibody (2I0Q4)
GREENER_CHOICE

Catalog No. PIMA542702
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIMA542702 100 μL
1 options
This item is not returnable. View return policy

Catalog No. PIMA542702

Supplier: Invitrogen™ MA542702

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive test controls include: A375. The target is usually found in the following locations: Chromosome, Nucleus. Immunogen sequence: AAAAGAAAAA AAEGEAPAEM GALLLEKETR GATERVHGSL GDTPRSEETL PKATPDSLEP AGPSSPASVT VTVGDEGADT PVGATPLIGD ESENLEGDGD LRGGRILLGH ATKSFPSSPS KGGSCPSRAK MSMTGAGKSP PSVQSLAMRL LSMPGAQGAA AAGSEPPPAT TSPEGQPKVH RARKTMSKPG NGQPPVPEKR PPEIQHFRMS DDVHSLGKVT SDLAKRRKLN SGGGLSEELG SARRSGEVTL TKGDPGSLEE WETVVGDDFS LYYDSYSVDE RVDSDSKSEV EALTEQLSE.

A cluster of genes, BAT1-BAT5, has been localized in the vicinity of the genes for TNF alpha and TNF beta. This gene is found near this cluster; it was mapped near the gene for C2 within a 120-kb region that included a HSP70 gene pair. These genes are all within the human major histocompatibility complex class III region. This gene was thought to be two different genes, NG36 and G9a, adjacent to each other but a recent publication shows that there is only a single gene. The protein encoded by this gene is thought to be involved in intracellular protein-protein interaction. There are three alternatively spliced transcript variants of this gene but only two are fully described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EHMT2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone 2I0Q4
Concentration 0.53 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Ehmt2
Gene Accession No. Q96KQ7, Q9Z148
Gene Alias ankyrin repeat-containing protein; Bat8; C6orf30; D17Ertd710e; DKFZp686H08213; Ehmt2; euchromatic histone lysine methyltransferase 2; euchromatic histone lysine N-methyltransferase 2; euchromatic histone-lysine N-methyltransferase 2; FLJ35547; G9a; G9A histone methyltransferase; GAT8; H3 ly; H3-K9-Histone-Methyl-Transferase; H3-K9-HMTase 3; histone H3-K9 methyltransferase 3; Histone-lysine N-methyltransferase EHMT2; histone-lysine N-methyltransferase, H3 lysine-9 specific 3; HLA-B associated transcript 8; HLA-B associated transcript 8, rat orthologue; HLA-B-associated transcript 8; KMT1C; lysine N-methyltransferase 1C; NG36; NG36/G9a; Protein G9a
Gene Symbols Ehmt2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 2-300 of human EHMT2 (Q96KQ7).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10919, 110147, 361798
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.