Learn More
STEMCELL Technologies EHNA (Hydrochloride), Size: 5 mg
SDP
Supplier:  STEMCELL Technologies 72442
Encompass_Preferred
EHNA is a reversible adenosine deaminase inhibitor (IC₅₀ = 1.2 μM in human red blood cells) that also selectively inhibits the cGMP-specific phosphodiesterase (PDE2) (IC₅₀ = 0.8 and 2 μM from human and porcine myocardium, respectively, 3.5 μM in rat hepatocyte, and 5.5 μM in human platelet) (Michie et al.; Podzuweit et al.). MAINTENANCE AND SELF-RENEWAL· Maintains pluripotency of human embryonic stem cells in the absence of exogenous cytokines, and blocks directed neuronal differentiation (Burton et al.).
Catalog No. 50-197-5923
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.