Learn More
Cayman Chemical EIcosapentaenoyl ChlorIde 10mg
SDP
Supplier:  Cayman Chemical 2272110 MG
Encompass_Preferred
A derivative of eicosapentaenoic acid; has been used in the synthesis of fatty acid conjugates to enhance lipophilicity and cell permeability of bioactive compounds
Catalog No. 50-299-5430