Learn More
Invitrogen™ EID1 Polyclonal Antibody

Description
The antibody detects endogenous levels of total EID1 protein.
Specifications
Specifications
| Antigen | EID1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.7 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | EID1 |
| Gene Accession No. | Q9DCR4, Q9Y6B2 |
| Gene Alias | 21 kDa pRb-associated protein; 2610002K20Rik; C15orf3; CREBBP/EP300 inhibitor 1; CREBBP/EP300 inhibitory protein 1; CRI1; E1A-like inhibitor of differentiation 1; EID; eid1; EID-1; EP300 interacting inhibitor of differentiation 1; EP300-interacting inhibitor of differentiation 1; IRO45620; NB4 apoptosis related protein; ORF12; PNAS-22; PTD014; Rb- and p300-binding protein EID-1; RBP21; retinoblastoma protein-associated protein; RGD1562702 |
| Gene Symbols | EID1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.