Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eIF1AX Recombinant Rabbit Monoclonal Antibody (1E8)
GREENER_CHOICE

Catalog No. MA572882
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA572882 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA572882 Supplier Invitrogen™ Supplier No. MA572882
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Recombinant Monoclonal Antibody

eIF1AX Recombinant Monoclonal Antibody for Flow, ICC/IF, IHC (F), IHC (P), IP, Western Blot

This gene encodes an essential eukaryotic translation initiation factor. The protein is required for the binding of the 43S complex (a 40S subunit, eIF2/GTP/Met-tRNAi and eIF3) to the 5' end of capped RNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF1AX
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 1E8
Concentration 0.3 mg/mL
Conjugate Unconjugated
Gene EIF1AX
Gene Accession No. P47813, Q8BMJ3
Gene Alias 1500010B24Rik; AI426898; EIF1A; eIF-1A; eIF-1A X isoform; EIF1AP1; EIF1AX; Eif1ay; EIF4C; eIF-4C; eukaryotic translation initiation factor 1A X-linked; eukaryotic translation initiation factor 1A, X chromosome; eukaryotic translation initiation factor 1A, X-chromosomal; eukaryotic translation initiation factor 1A, X-linked; eukaryotic translation initiation factor 1A, Y-linked; eukaryotic translation initiation factor 4C; Putative eukaryotic translation initiation factor 1A; RGD1560198; X-linked eukaryotic translation initiation factor 1A
Gene Symbols EIF1AX
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human eIF1AX
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1964, 302697, 66235
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.