Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eIF2 gamma Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA592081
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA592081 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA592081 Supplier Invitrogen™ Supplier No. PA592081
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HLAAIEIMKL KHILILQNKI DLVKESQAKE QYEQILAFVQ GTVAEGAPII PISAQLKYNI EVVCEYIVKK IPVPPRDFTS EPRLIVIRSF DVNKPGCEVD DLKGGVAGGS ILKGVLKVGQ EIEVRPGIVS KDSEGKLMCK PIFSKIVSLF AEHNDLQYAA PGGLIGVGTK IDPTLCRADR MVGQVLGAVG ALPEIFTELE ISYFLLRRLL GVRTEGDKKA AKVQKLSKNE VLMVNIGSLS TGGRVSAVKA DLGKIVLTNP VCTEVGEKIA LSRRVEKHWR LIGWGQIRRG VTIKPTVDDD; Positive Samples: Rat brain.

The initiation of protein synthesis in eukaryotic cells is regulated by interactions between protein initiation factors and RNA molecules. The eukaryotic initiation complex is composed of three subunits, designated eIF2 alpha, eIF2 beta and eIF2 gamma (eukaryotic translation initiation factor 2 alpha, beta and gamma, respectively), all of which work in concert to form a ternary complex with GTP and tRNA in the early stages of protein synthesis. eIF2S3 (eukaryotic translation initiation factor 2, subunit 3), also known as eIF2G, is a 472 amino acid protein that belongs to the gamma subfamily of GTP-binding elongation factor proteins. Existing as a heterotrimer composed of an alpha, beta and gamma chain, eIF2S3 functions to bind ribosomal subunits and catalyze the subsequent formation of preinitiation complexes necessary for protein synthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF2 gamma
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.26 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene EIF2S3
Gene Accession No. P81795
Gene Alias EIF2; EIF2G; eIF-2gA; EIF2gamma; eIF-2-gamma; eIF-2-gamma X; eIF-2gX; Eif2s3; Eif2s3x; Eukaryotic translation initiation factor 2 subunit 3; Eukaryotic translation initiation factor 2 subunit 3, X-linked; Eukaryotic translation initiation factor 2 subunit gamma; eukaryotic translation initiation factor 2 subunit gamma X; Eukaryotic translation initiation factor 2 subunit gamma, X-linked; eukaryotic translation initiation factor 2, subunit 3 gamma, 52kDa; eukaryotic translation initiation factor 2, subunit 3, structural gene X-linked; eukaryotic translation initiation factor 2G; PP42
Gene Symbols EIF2S3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 173-472 of human EIF2S3 (NP_0014061).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 299027
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.