Learn More
EIF4E2 Mouse anti-Human, Clone: OTI1F5, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF502402
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Recognizes and binds the 7-methylguanosine-containing mRNA cap during an early step in the initiation of protein synthesis and facilitates ribosome binding by inducing the unwinding of the mRNAs secondary structures.Specifications
| EIF4E2 | |
| Monoclonal | |
| Unconjugated | |
| EIF4E2 | |
| 4E-LP, 4EHP, EIF4EL3, IF4e, h4EHP | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9470 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI1F5 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| O60573 | |
| EIF4E2 | |
| Full length human recombit protein of human EIF4E2 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.