Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ EIF4E2 Recombinant Protein Antigen
SDP

Catalog No. NB171583 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB171583 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB171583 Supplier Novus Biologicals™ Supplier No. NBP317839PEP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Specifications

For Use With (Application) Antibody Competition
Formulation PBS and 1M Urea, pH 7.4.
Gene ID (Entrez) 9470
Molecular Weight (g/mol) M.W. Theoretical: 24 kDa
Quantity 100 μg
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Gene Alias 4EHP, 4E-LP, eIF4E type 2, eIF-4E type 2, EIF4EL3, eIF4E-like cap-binding protein, eIF4E-like protein 4E-LP, eukaryotic translation initiation factor 4E family member 2, Eukaryotic translation initiation factor 4E homologous protein, eukaryotic translation initiation factor 4E member 2, eukaryotic translation initiation factor 4E type 2, Eukaryotic translation initiation factor 4E-like 34E-LP, IF4e, mRNA cap-binding protein 4EHP, mRNA cap-binding protein type 3
Gene Symbol EIF4E2
Research Category DNA Repair, DNA replication Transcription Translation and Splicing
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein EIF4E2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.