Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eIF5A2 Monoclonal Antibody (OTI6G7)
GREENER_CHOICE

Catalog No. PIMA526047
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIMA526047 100 μL
1 options

Catalog No. PIMA526047

Supplier: Invitrogen™ MA526047

Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

eIF5A2 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

mRNA-binding protein involved in translation elongation. Has an important function at the level of mRNA turnover, probably acting downstream of decapping. Involved in actin dynamics and cell cycle progression, mRNA decay and probably in a pathway involved in stress response and maintenance of cell wall integrity. Functions as a regulator of apoptosis. Mediates effects of polyamines on neuronal process extension and survival. May play an important role in brain development and function, and in skeletal muscle stem cell differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF5A2
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI6G7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene EIF5A2
Gene Accession No. Q9GZV4
Gene Alias 9630038B20; EIF5A2; eIF-5A2; eIF-5A-2; eIF5AII; eukaryotic initiation factor 5A; Eukaryotic initiation factor 5A isoform 2; eukaryotic translation initiation factor 5A2; Eukaryotic translation initiation factor 5A-2
Gene Symbols EIF5A2
Host Species Mouse
Immunogen Full length human recombinant protein of EIF5A2 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56648
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.