Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eIF5B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590237
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590237 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590237 Supplier Thermo Scientific Supplier No. PA590237
Only null left
Add to Cart
Add to Cart

eIF5B Polyclonal Antibody

eIF5B Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF5B
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.84 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene EIF5B
Gene Accession No. B2GUV7, O60841, Q05D44
Gene Alias A030003E17Rik; Annexin V-binding protein ABP-7; BC018347; DKFZp434I036; eIF 5B; Eif5b; eIF-5B; Eukaryotic translation initiation factor 5B; FLJ10524; If2; KIAA0741; translation initiation factor IF2; Translation initiation factor IF-2
Gene Symbols EIF5B
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-273 of human EIF5B (NP_0569883).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 226982, 308306, 9669
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.