Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eIF6 Monoclonal Antibody (2I11)
GREENER_CHOICE

Catalog No. PIMA549259
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549259 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549259 Supplier Invitrogen™ Supplier No. MA549259
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Human EIF6 shares 99.3% amino acid (aa) sequence identity with both mouse and rat EIF6. Positive Control - WB: human placenta tissue, human CACO-2 whole cell, human HEPG2 whole cell, human K562 whole cell, human A549 whole cell, human PC-3 whole cell, human Hela whole cell, human U87 whole cell, rat kidney tissue, rat testis tissue, mouse HEPA1-6 whole cell. IHC: human intestinal cancer tissue, human mammary cancer tissue, human lung cancer tissue, rat spleen tissue. Flow: U20S cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Hemidesmosomes are structures which link the basal lamina to the intermediate filament cytoskeleton. An important functional component of hemidesmosomes is the integrin beta-4 subunit (ITGB4), a protein containing two fibronectin type III domains. The protein encoded by this gene binds to the fibronectin type III domains of ITGB4 and may help link ITGB4 to the intermediate filament cytoskeleton. The encoded protein, which is insoluble and found both in the nucleus and in the cytoplasm, can function as a translation initiation factor and prevent the association of the 40S and 60S ribosomal subunits. Multiple transcript variants encoding two different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF6
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 2I11
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.05mg sodium azide
Gene Eif6
Gene Accession No. O55135, P56537, Q3KRD8
Gene Alias 1110004P11; AA408895; b(2)gcn; B(2)GCN homolog; B4 integrin interactor; CAB; EIF3A; Eif6; eIF-6; eIF-6 {ECO:0000255; eukaryotic translation initiation factor 3A; eukaryotic translation initiation factor 6; eukaryotic translation initiation factor 6 {ECO:0000255; HAMAP-Rule:MF_03132}; imc-415; integrin beta 4 binding protein; Itgb4bp; OK/SW-cl.27; p27 beta-4 integrin-binding protein; p27(BBP); p27BBP
Gene Symbols Eif6
Host Species Mouse
Immunogen E.coli-derived human EIF6 recombinant protein (Position: N66-T210).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16418, 305506, 3692
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.